Chloroform-[D₁] 99.8+ atom % D + 0.02-0.04 % v/v TMS for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

464020075
TS464020075EA 36 USD
TS464020075
Chloroform-[D₁] 99.8+ atom % D + 0.02-0.04 % v/v TMS for NMR spectroscopy
Chloroform-[D1]
Formula: CDCl₃
MW: 120.38 g/mol
MDL Number: MFCD00000827
CAS Number: 865-49-6
UN: 1888

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR