Nobiletin

Supplier: Thermo Scientific Chemicals

Synonyms: 3',4',5,6,7,8-Hexamethoxyflavone, 2-(3,4-Dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-chromen-4-one

46352-2500
TS46352-0010EA 466 USD
TS46352-0010 TS46352-2500
Nobiletin
Nobiletin
Formula: C₂₁H₂₂O₈
MW: 402.40 g/mol
Melting Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03273560
CAS Number: 478-01-3

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
Water ≤5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR