Gemfibrozil 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid, CI-719, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeric acid
46179-0050
46179-0250
TS46179-0050EA
74
USD
TS46179-0050
TS46179-0250
Gemfibrozil 98%
Gemfibrozil
Formula:
C₁₅H₂₂O₃ MW: 250.34 Boiling Pt: 395 Melting Pt: 60…61 °C Storage Temperature: Freezer |
MDL Number:
MFCD00079335 CAS Number: 25812-30-0 Merck Index: 12,04394 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 58 to 63 °C |
Water | ≤0.25 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...