1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulfonyl)-β-D-mannopyranose, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulfonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, TATM

46134-0010 46134-1000
TS46134-0010EA 416 USD
TS46134-0010 TS46134-1000
1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose 98%
Mannose triflate
Formula: C₁₅H₁₉F₃O₁₂S
Melting Pt: 118…122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012353
CAS Number: 92051-23-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 118 to 122 °C
Solubility Clear colorless to slightly yellow
HPLC ≥97.5 % (ELSD)
Specific optical rotation -12° to -19° (20 °C, 589 nm) (1 % in CH3CN)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR