Lamivudine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (-)-1-[(2R,5S)-2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]cytosine, 4-Amino-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one, 3TC

46109-0010
TS46109-0010EA 154 USD
TS46109-0010
Lamivudine 98%
Lamivudine
Formula: C₈H₁₁N₃O₃S
MW: 229.2600 g/mol
Melting Pt: 160…162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00869739
CAS Number: 134678-17-4

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 175 to 179 °C
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR