Copper (II) ethylenediamine 1 M in water, Technical Grade not for viscosity determination

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(ethylenediamine)copper dihydroxide, Cupriethylendiamine, Copper(II)-ethylenediame complex

46068-0010
TS46068-0010EA 41 USD
TS46068-0010
Copper (II) ethylenediamine 1 M in water, Technical Grade not for viscosity determination
Copper (II) ethylenediamine
Formula: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MW: 217.76 g/mol
Density: 1.1 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00274628
CAS Number: 14552-35-3
UN: 1761
ADR: 8,II

Order Now

Specification Test Results

Appearance Dark blue Solution
Strength 0.98 to 1.02 M (copper molarity)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR