Methyl nervonate 99% analytical standard for GC

Supplier: Thermo Scientific Chemicals

46037-1000 46037-5000
TS46037-1000EA 93 USD
TS46037-1000 TS46037-5000
Methyl nervonate 99% analytical standard for GC
Methyl nervonate
Formula: C₂₅H₄₈O₂
Storage Temperature: Freezer
MDL Number: MFCD00056324
CAS Number: 2733-88-2

Order Now

Specification Test Results

Appearance White low melting solid
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR