Sodium dodecyl sulfate (SDS), granular, pure, meets analytical specification of Ph.Eur.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

AC45900-0010 AC45900-0050
TS45900-0010EA 63 USD
TS45900-0010 TS45900-0050
Sodium dodecyl sulfate (SDS), granular, pure, meets analytical specification of Ph.Eur.
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 206 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Identification A Passes test
Identification B Passes test
Identification C Passes test
Identification D Passes test
Alkalinity Passes test
Titration ≥85 %
NaCl and Na2SO4 ≤8 %
Non-esterified alcohols ≤4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR