Voriconazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2R,3S)-2-(2,4-Difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol, (2R,3S)-2-(2,4-Difluorophenyl)-3-(5-fluoro-4-pyrimidinyl)-1-(1H-1,2,4-triazol-1-yl)-2-butanol

45867-0010
TS45867-0010EA 191 USD
TS45867-0010
Voriconazole 98%
Voriconazole
Formula: C₁₆H₁₄F₃N₅O
MW: 349.32 g/mol
Melting Pt: 128…134 °C
Storage Temperature: Freezer
MDL Number: MFCD00905717
CAS Number: 137234-62-9
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR