Polyvinyl alcohol, M.W. ∼ 124,000 99-100%, hydrolysed
Supplier: Thermo Scientific Chemicals
Synonyms:
Poly(vinyl alcohol), PVOH, PVA, Mowiol®
458230250
458230010
458230050
TS458230250EA
37
USD
TS458230250
TS458230010
TS458230050
Polyvinyl alcohol, M.W. ∼ 124,000 99-100%, hydrolysed
Polyvinyl alcohol
Formula:
[-CH₂CH(OH)-]n Melting Pt: 230…240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081922 CAS Number: 9002-89-5 |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Viscosity | 28 to 32 cP (4 % in water) |
Degree of hydrolysis | 99.3 to 100.0 mol % |
Ash | ≤1.2 % (as Na2O) |
pH | 5.5 to 7.5 (4 % in water) |
Volatile matter | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...