Magnesium silicate 60-100 mesh for pesticide residue analysis

Supplier: Thermo Scientific Chemicals

457725000
TS457725000EA 230 USD
TS457725000
Magnesium silicate 60-100 mesh for pesticide residue analysis
Magnesium silicate
Formula: MgO₃Si
Storage Temperature: Ambient
MDL Number: MFCD00078249
CAS Number: 1343-88-0

Order Now

Specification Test Results

Appearance White Powder (May contain black particles)
Silicate (as SiO2) 83.5 to 84.5 %
Sodium sulfate (Na2SO4) ≤1.0 %
Magnesium oxide (MgO) 15 to 16 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR