2-Chlorothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

45726-0010 45726-0050 45726-0250
TS45726-0010EA 40 USD
TS45726-0010 TS45726-0050 TS45726-0250
2-Chlorothiazole 97%
2-Chlorothiazole
Formula: C₃H₂ClNS
Boiling Pt: 145…146 °C (760 mmHg)
Density: 1.378 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00210701
CAS Number: 3034-52-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR