Inulin (from chicory root)

Supplier: Thermo Scientific Chemicals

45710-0250 45710-1000 45710-5000
TS45710-0250EA 136 USD
TS45710-0250 TS45710-1000 TS45710-5000
Inulin (from chicory root)
Inulin
Storage Temperature: Ambient
MDL Number: MFCD00131407
CAS Number: 9005-80-5

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (2 g, 105°C, 2 hrs)
Free fructose, glucose,sucrose ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR