Inulin (from chicory root)
Supplier: Thermo Scientific Chemicals
45710-0250
45710-1000
45710-5000
TS45710-0250EA
136
USD
TS45710-0250
TS45710-1000
TS45710-5000
Inulin (from chicory root)
Inulin
Storage Temperature:
Ambient |
MDL Number:
MFCD00131407 CAS Number: 9005-80-5 |
Specification Test Results
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤10 % (2 g, 105°C, 2 hrs) |
Free fructose, glucose,sucrose | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...