5-Bromo-1H-indazole 97%

Supplier: Thermo Scientific Chemicals
Danger

45674-0250 45674-0050
TS45674-0250EA 282 USD
TS45674-0250 TS45674-0050
5-Bromo-1H-indazole 97%
5-Bromo-1H-indazole
Formula: C₇H₅BrN₂
Melting Pt: 123…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00839493
CAS Number: 53857-57-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 125 to 132 °C
Water ≤1.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR