7-Azaindole-5-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

45585-0010
TS45585-0010EA 204 USD
TS45585-0010
7-Azaindole-5-boronic acid pinacol ester 97%
7-Azaindole-5-boronic acid pinacol ester
Formula: C₁₃H₁₇BN₂O₂
Melting Pt: 243…245.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08060937
CAS Number: 754214-56-7

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 243 to 249 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR