Cefoxitin sodium 94%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Mefoxin, Cefoxitin sodium salt, Sodium (6R,7S)-3-((carbamoyloxy)methyl)-7-methoxy-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

45528-0010 45528-0050
TS45528-0010EA 138 USD
TS45528-0010 TS45528-0050
Cefoxitin sodium 94%
Cefoxitin sodium
Formula: C₁₆H₁₆N₃NaO₇S₂
MW: 449.428 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00079042
CAS Number: 33564-30-6

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥93 %
Specific optical rotation +206° to +214° (20°C, 589 nm) (c= 1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR