Methyl arachidate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl eicosanoate, Methyl Arachidate
45282-0010
45282-0050
45282-0250
TS45282-0010EA
84
USD
TS45282-0010
TS45282-0050
TS45282-0250
Methyl arachidate 99%
Methyl arachidate
Formula:
C₂₁H₄₂O₂ Boiling Pt: 215…216 °C (10 mmHg) Melting Pt: 45…48 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009014 CAS Number: 1120-28-1 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 45 to 48 °C |
GC | ≥98.5 % |
Free acid | ≤1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...