1-Boc-azetidine-3-ylmethanol 96%

Supplier: Thermo Scientific Chemicals
Warning

45209-0050
TS45209-0050EA 408 USD
TS45209-0050
1-Boc-azetidine-3-ylmethanol 96%
1-Boc-azetidine-3-ylmethanol
Formula: C₉H₁₇NO₃
Melting Pt: 50…53 °C
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD06656141
CAS Number: 142253-56-3

Order Now

Specification Test Results

Appearance Colorless to white to yellow Crystals or low melting solid
Infrared spectrum Conforms
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR