L-Alanine 99%, meets analytical specification of Ph.Eur.

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

45185-0010 45185-0025 45185-2500
TS45185-0010EA 175 USD
TS45185-0010 TS45185-0025 TS45185-2500
L-Alanine 99%, meets analytical specification of Ph.Eur.
L-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189
Melting Pt: 79 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
Merck Index: 13,00203

Order Now

Specification Test Results

Appearance Colorless to white to almost white Crystals or crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Sulfated ash ≤0.1 % (1 g, 600°C)
Appearance of solution Clear and not more intensely colored than reference solution BY6
Specific optical rotation +13.5° to +15.5° (20°C, 589 nm) (c=10, 6 N HCl) (dried substance)
Titration with HClO4 98.5 to 101.0 % (dried substance)
Sulfates ≤300 ppm
Chlorides ≤200 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤10 ppm
Ninhydrin-positive substances ≤0.5 % (Total)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR