Tin(IV) oxide 99%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

AC45181-0010 AC45181-0025 AC45181-1000
TS45181-0010EA 131 USD
TS45181-0010 TS45181-0025 TS45181-1000
Tin(IV) oxide 99%, pure
Tin (IV) oxide
Formula: O₂Sn
MW: 150.71 g/mol
Melting Pt: 1624.85
Density: 6.95 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011244
CAS Number: 18282-10-5
Merck Index: 14,08778

Order Now

Specification Test Results

Appearance White to light gray Powder
Purity ≥98.5 % (metals basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR