Sodium hydroxide, micropearls, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

45113-0050 45113-0250
TS45113-0050EA 53 USD
TS45113-0050 TS45113-0250
Sodium hydroxide, micropearls, extra pure
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Appearance White Pearls
Particle size 20 to 40 mesh
Titration with HCl ≤2.5 % (Na2CO3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR