2,2,3,3-Tetramethoxybutane 95%

Supplier: Thermo Scientific Chemicals
Warning

45093-0050 45093-0250
TS45093-0050EA 110 USD
TS45093-0050 TS45093-0250
2,2,3,3-Tetramethoxybutane 95%
2,2,3,3-Tetramethoxybutane
Formula: C₈H₁₈O₄
Boiling Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD25458126
CAS Number: 176798-33-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR