D-(+)-Glucose monohydrate 99+%, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate

45074-0010 45074-0050 45074-0250
TS45074-0010EA 41 USD
TS45074-0010 TS45074-0050 TS45074-0250
D-(+)-Glucose monohydrate 99+%, extra pure
D(+)-Glucose monohydrate
Formula: C₆H₁₂O₆·H₂O
MW: 198.17 g/mol
Melting Pt: 83
Storage Temperature: Ambient
MDL Number: MFCD00149450
CAS Number: 14431-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 8 to 10 %
HPLC ≥99.0 % (ELSD)
Residue after ign. (sulfated) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR