D-(+)-Glucose monohydrate 99+%, extra pure
Supplier: Thermo Scientific Chemicals
Synonyms:
(2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
45074-0010
45074-0050
45074-0250
TS45074-0010EA
41
USD
TS45074-0010
TS45074-0050
TS45074-0250
D-(+)-Glucose monohydrate 99+%, extra pure
D(+)-Glucose monohydrate
Formula:
C₆H₁₂O₆·H₂O MW: 198.17 g/mol Melting Pt: 83 Storage Temperature: Ambient |
MDL Number:
MFCD00149450 CAS Number: 14431-43-7 |
|
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | 8 to 10 % |
HPLC | ≥99.0 % (ELSD) |
Residue after ign. (sulfated) | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...