1,4-Dioxane, extra dry 99.8% stabilized, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

450171000
TS450171000EA 69 USD
TS450171000
1,4-Dioxane, extra dry 99.8% stabilized, AcroSeal®
1,4-Dioxane
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: 12 °C
Density: 1.034 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
UN: 1165
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.75 %
Stabilizer 2 to 5 ppm BHT
Residue after evaporation ≤0.001 %
Acidity (CH3COOH) ≤0.002
Peroxides ≤0.005 % (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR