Polyvinyl alcohol, M.W. ∼ 85,000-120,000 88%, hydrolysed
Supplier: Thermo Scientific Chemicals
Synonyms:
Poly(vinyl alcohol), PVOH, PVA, Mowiol®
450140010
450140250
450145000
TS450140010EA
170
USD
TS450140010
TS450140250
TS450145000
Polyvinyl alcohol, M.W. ∼ 85,000-120,000 88%, hydrolysed
Polyvinyl alcohol
Formula:
[-CH₂CH(OH)-]n Melting Pt: 230…240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081922 CAS Number: 9002-89-5 |
Specification Test Results
Appearance | White to off-white Granular solid |
Infrared spectrum | Conforms |
Viscosity | 23.0 to 27.0 cP (4 % in water) |
Degree of hydrolysis | 87.0 to 89.0 mol % |
Ash | ≤0.5 % (Na2O) |
pH | 4.5 to 6.5 (4 % in water) |
Volatile matter | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...