Perfluorokerosene 50% in PFK low boiling, high boiling for mass spectrometry

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: PFK

45013-0010
TS45013-0010EA 336 USD
TS45013-0010
Perfluorokerosene 50% in PFK low boiling, high boiling for mass spectrometry
Perfluorokerosene
Boiling Pt: 180 °C
Density: 1.89 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00132835
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Suitability (for mass analysis) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR