2',3'-Dideoxycytidine 98%

Warning

Synonyms: 4-Amino-1-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Zalcitabine

44950-1000
TS44950-1000EA 103 USD
TS44950-1000 TS44950-5000
2',3'-Dideoxycytidine 98%
2',3'-Dideoxycytidine
Formula: C₉H₁₃N₃O₃
MW: 211.22 g/mol
Melting Pt: 216…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012188
CAS Number: 7481-89-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR