Methyl hexanoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester

44924-0010 44924-0500 44924-2500
TS44924-0010EA 164 USD
TS44924-0010 TS44924-0500 TS44924-2500
Methyl hexanoate 99%
Methyl hexanoate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 151
Melting Pt: -63
Density: 0.884 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00009510
CAS Number: 106-70-7
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR