(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 98%

Supplier: Thermo Scientific Chemicals
Warning

44921-0250 44921-0050
TS44921-0250EA 92 USD
TS44921-0250 TS44921-0050
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 98%
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione
Formula: C₆H₈O₄
Boiling Pt: 255 °C (760 mmHg)
Melting Pt: 95…98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00070594
CAS Number: 4511-42-6

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation -275° to -295° (20°C, 589 nm) (c=1, toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR