(-)-Epigallocatechin (from green tea) 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,3R)-2-(3,4,5-Trihydroxyphenyl)chromane-3,5,7-triol, 3,5,7,3',4',5'-Hexahydroxylflavan

44898-0100
TS44898-0100EA 201 USD
TS44898-0100
(-)-Epigallocatechin (from green tea) 95%
(-)-Epigallocatechin
Formula: C₁₅H₁₄O₇
MW: 306.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075939
CAS Number: 970-74-1

Order Now

Specification Test Results

Appearance White to light pink to light brown Powder
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR