1-Bromo-3-chloro-5-fluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloro-5-fluorobromobenzene

448750250
TS448750250EA 162 USD
TS448750250
1-Bromo-3-chloro-5-fluorobenzene 98%
1-Bromo-3-chloro-5-fluorobenzene
Formula: C₆H₃BrClF
Boiling Pt: 72 °C (5 mmHg)
Density: 1.72 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00070747
CAS Number: 33863-76-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5470 to 1.5490 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR