Sodium borohydride 99%, caplets, VenPure™ AF, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

44849-5000 AC44849-0100 AC44849-1000
TS44849-5000EA 304 USD
TS44849-5000 TS44849-0100 TS44849-1000
Sodium borohydride 99%, caplets, VenPure™ AF, pure
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400 (dec.)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664

Order Now

Specification Test Results

Appearance White Free flowing caplets
Titration with Na2S2O3 ≥98.5 %
Silicate (as SiO2) ≤50 ppm
Magnesium carbonate ≤100 ppm
Iron (Fe) ≤5 ppm
Fines ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR