Tetrahydrofuran, extra dry 99.85% stabilized, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC44836-1000
TS44836-1000EA 81 USD
TS44836-1000
Tetrahydrofuran, extra dry 99.85% stabilized, AcroSeal™
Tetrahydrofuran
Formula: C₄H₈O
Boiling Pt: 66 °C (760 mmHg)
Melting Pt: –108.4 °C
Density: 0.88 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
UN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.005 % (Coulometric)
GC ≥99.80 %
Stabilizer ≥200 ppm BHT
Residue after evaporation ≤0.001 %
Filt.over 0.2 micron PFTE Filtered
Acidity (CH3COOH) ≤0.0005 meq/g
Peroxides ≤0.0025 % (H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR