Bismuth sodium trioxide ACS

Supplier: Thermo Scientific Chemicals
Warning

AC44813-0250 AC44813-1000
TS44813-0250EA 100 USD
TS44813-0250 TS44813-1000
Bismuth sodium trioxide ACS
Bismuth sodium trioxide
Formula: BiNaO₃
Storage Temperature: Ambient
MDL Number: MFCD00003474
CAS Number: 12232-99-4

Order Now

Specification Test Results

Appearance Yellow to brown Solid
Titration with KMnO4 ≥80.0 %
Chloride (Cl) ≤0.002 %
Manganese (Mn) ≤5 ppm
Oxidizing efficiency ≥99.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR