Mercury(II) bromide ACS

Supplier: Thermo Scientific Chemicals
Danger

AC44812-1000
TS44812-1000EA 107 USD
TS44812-1000
Mercury(II) bromide ACS
Mercury (II) bromide
Formula: Br₂Hg
Boiling Pt: 322 °C (760 mmHg)
Melting Pt: 236 °C
Storage Temperature: Ambient
MDL Number: MFCD00011040
CAS Number: 7789-47-1
UN: 1634
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to very light yellow Powder
Titration Complexometric ≥98.0 %
Chloride (Cl) ≤0.25 %
Insoluble matter ≤0.05 % (In methanol)
Residue after reduction ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR