Pyridine 99.8%, AcroSeal® for biochemistry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Azabenzene, Azine

447900010
TS447900010EA 161 USD
TS447900010
Pyridine 99.8%, AcroSeal® for biochemistry
Pyridine
Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115…116 °C (760 mmHg)
Melting Pt: –42 °C
Density: 0.978 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
UN: 1282
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.75 %
UV at 370 nm A: ≤0.004
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Chloride (Cl) ≤0.0005 %
Sulfate (SO4) ≤0.0005 %
Cadmium (Cd) ≤0.000005 %
Copper (Cu) ≤0.000002 %
Ammonia (NH3) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR