Potassium nitrate 99+% for biochemistry

Supplier: Thermo Scientific Chemicals
Warning

44782-0025 44782-5000
TS44782-0025EA 302 USD
TS44782-0025 TS44782-5000
Potassium nitrate 99+% for biochemistry
Potassium nitrate
Formula: KNO₃
Boiling Pt: 400 °C (760 mmHg)
Melting Pt: 334 °C
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
UN: 1486
ADR: 5.1,III

Order Now

Specification Test Results

Appearance White Crystals
Solubility (1 M in water) Clear colorless
pH 4.5 to 8.5
Titration after Ion exchange ≥99.0 %
Chloride (Cl) ≤0.002 %
Nitrite (NO2) ≤0.001 %
Phosphate (PO4) ≤0.003 %
Sulfate (SO4) ≤0.015 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR