Lead(II) perchlorate trihydrate ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
447781000
447785000
TS447781000EA
83
USD
TS447781000
TS447785000
Lead(II) perchlorate trihydrate ACS
Lead (II) perchlorate trihydrate
Formula:
Cl₂O₈Pb·3H₂O Melting Pt: 100 °C (dec.) Density: 2.6 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149817 CAS Number: 13453-62-8 UN: 1470 ADR: 5.1,II |
Specification Test Results
Appearance | White Crystalline powder |
Titration Complexometric | 97.0 to 102.0 % |
pH | 3.0 to 5.0 (5% in water, 25.0°C) |
Chloride (Cl) | ≤0.01 % |
Iron (Fe) | ≤0.001 % |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...