Potassium carbonate, anhydrous 98%, extra pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potash, Carbonic acid potassium salt, Carbonic acid, dipotassium salt

AC44762-0010 AC44762-0050 AC44762-0250
TS44762-0010EA 48 USD
TS44762-0010 TS44762-0050 TS44762-0250
Potassium carbonate, anhydrous 98%, extra pure
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying ≤2.0 %
Titration with HClO4 ≥97.5 %
Heavy metals (as Pb) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR