Magnesium nitrate hexahydrate, extra pure
Supplier: Thermo Scientific Chemicals
AC44754-0010
AC44754-0050
AC44754-0250
TS44754-0010EA
55
USD
TS44754-0010
TS44754-0050
TS44754-0250
Magnesium nitrate hexahydrate, extra pure
Magnesium nitrate hexahydrate
Formula:
Mg(NO₃)₂·6H₂O Boiling Pt: 330 °C (760 mmHg) Melting Pt: 89 °C Flash Pt: 93.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 |
Specification Test Results
Appearance | White Crystals |
Titration Complexometric | 98 to 102 % |
pH | 5 to 8.2 (5 % in water) |
Chloride (Cl) | ≤0.001 % |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤0.001 % |
Heavy metals (as Pb) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...