Magnesium nitrate hexahydrate, extra pure

Supplier: Thermo Scientific Chemicals

AC44754-0010 AC44754-0050 AC44754-0250
TS44754-0010EA 55 USD
TS44754-0010 TS44754-0050 TS44754-0250
Magnesium nitrate hexahydrate, extra pure
Magnesium nitrate hexahydrate
Formula: Mg(NO₃)₂·6H₂O
Boiling Pt: 330 °C (760 mmHg)
Melting Pt: 89 °C
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric 98 to 102 %
pH 5 to 8.2 (5 % in water)
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR