Sodium perchlorate, anhydrous ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
44742-0025
44742-5000
AC44742-1000
TS44742-0025EA
602
USD
TS44742-0025
TS44742-5000
TS44742-1000
Sodium perchlorate, anhydrous ACS
Sodium perchlorate
Formula:
ClNaO₄ Melting Pt: 482 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003480 CAS Number: 7601-89-0 UN: 1502 ADR: 5.1,II |
Specification Test Results
Appearance | White Powder or crystals and/or chunks |
Titration Argentimetric | 98.0 to 102.0 % |
pH | 6.0 to 8.0 (5% solution at 25.0°C) |
Chloride (Cl) | ≤0.003 % |
Sulfate (SO4) | ≤0.002 % |
Calcium (Ca) | ≤0.02 % |
Iron (Fe) | ≤5 ppm |
Potassium (K) | ≤0.05 % |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...