Molybdenum(VI) oxide 99.5+% ACS

Supplier: Thermo Scientific Chemicals
Warning

AC44740-1000 AC44740-5000
TS44740-1000EA 84 USD
TS44740-1000 TS44740-5000
Molybdenum(VI) oxide 99.5+% ACS
Molybdenum (VI) oxide
Formula: MoO₃
Boiling Pt: 1155 °C (760 mmHg)
Melting Pt: 795 °C
Storage Temperature: Ambient
MDL Number: MFCD00003469
CAS Number: 1313-27-5

Order Now

Specification Test Results

Appearance White to yellow to green to gray Powder
Titration ≥99.5 % (Precipitation titration of molybdate)
Chloride (Cl) ≤0.002 %
Nitrate (NO3) Passes test (Limit about 0.003%)
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.02 %
AsO4, PO4, SiO4 (as SiO2) ≤0.001 %
Ammonium (NH4) ≤0.002 %
Heavy metals (as Pb) ≤0.005 %
Insoluble matter ≤0.01 % (In dilute NH4OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR