di-Potassium hydrogen phosphate, anhydrous 99% for HPLC

Supplier: Thermo Scientific Chemicals

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

44736-2500
TS44736-2500EA 280 USD
TS44736-2500
di-Potassium hydrogen phosphate, anhydrous 99% for HPLC
di-Potassium hydrogen phosphate
Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340 °C
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or granules
Loss on drying ≤1 %
Solubility Clear colorless
UV at 500 nm A: ≤0.02
pH 8.5 to 9.6 (10 % in water)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR