Calcium chloride dihydrate 99% for biochemistry

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

AC44732-0025 AC44732-5000
TS44732-0025EA 254 USD
TS44732-0025 TS44732-5000
Calcium chloride dihydrate 99% for biochemistry
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600
Melting Pt: 176
Density: 0.83 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
Merck Index: 13,01660

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric ≥98.5 %
Solubility Clear colorless
Acidity Passes test
Alkalinity Passes test
Sulfate (SO4) ≤0.03 %
Aluminium (Al) ≤1 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR