Mercury(II) acetate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

AC44727-0250 AC44727-1000 AC44727-5000
TS44727-0250EA 62 USD
TS44727-0250 TS44727-1000 TS44727-5000
Mercury(II) acetate ACS
Mercury (II) acetate
Formula: (H₃CCOO)₂Hg
Melting Pt: 179…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
UN: 1629
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White Powder
Titration with NH4SCN ≥98.0 %
Chloride (Cl) ≤0.005 %
Nitrate (NO3) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Other heavy metals (as Pb) ≤0.002 %
Mercurous Mercury (as Hg) ≤0.4 %
Insoluble matter ≤0.01 %
Residue after reduction ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR