Potassium chromate ACS, meets analytical specifications of Reagent Ph. Eur.

Supplier: Thermo Scientific Chemicals
Danger

AC44720-1000 AC44720-5000
TS44720-1000EA 62 USD
TS44720-1000 TS44720-5000
Potassium chromate ACS, meets analytical specifications of Reagent Ph. Eur.
Potassium chromate
Formula: K₂CrO₄
MW: 194.19 g/mol
Boiling Pt: 1000
Melting Pt: 985
Storage Temperature: Ambient
MDL Number: MFCD00011368
CAS Number: 7789-00-6
UN: 3077
ADR: 9 III
Merck Index: 12,07784

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Titration Iodometric ≥99.0 %
pH 8.6 to 9.8 (5% soln. at 25°C)
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.03 %
Calcium (Ca) ≤0.005 %
Sodium (Na) ≤0.02 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR