Carbon disulfide 99.9% ACS

Supplier: Thermo Scientific Chemicals
Danger

44703-5000
TS44703-5000EA 74 USD
TS44703-5000
Carbon disulfide 99.9% ACS
Carbon disulphide
Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46 °C (760 mmHg)
Melting Pt: –111 °C
Density: 1.262 g/cm³
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
UN: 1131
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 %
GC ≥99.9 %
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Sulfur dioxide (SO2) ≤2.5 ppm
Hydrogen sulfide (H2S) ≤1.5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR